5-Amino-1H-imidazole-4-carboxamide hydrochloride 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
5(4)-Amino-4(5)-imidazolecarboxamide Hydrochloride, AICA, 5(4)-Amino-4(5)-(aminocarbonyl)imidazole Hydrochloride, 4-Aminoimidazole-5-carboxamide hydrochloride, 4-Amino-5-imidazolecarboxamide hydrochloride, 5-Amino-4-imidazolecarboxamide hydrochloride
17309-0050
17309-0250
TS17309-0050EA
73
USD
TS17309-0050
TS17309-0250
5-Amino-1H-imidazole-4-carboxamide hydrochloride 98%
5-Amino-1H-imidazole-4-carboxamide hydrochloride
Formula:
C₄H₇ClN₄O MW: 162.58 g/mol Melting Pt: 250 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012704 CAS Number: 72-40-2 |
Specification Test Results
Appearance | White to light beige to light grey Powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥97.5 % |
Solubility | (5 % in water) Clear |
Residue after evaporation | ≤0.001 % |
Acidity | ≤0.0005 meq/g |
Peroxides | ≤0.0075 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...