4-Carbamoylphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

44490-0050 44490-0250
TS44490-0050EA 163 USD
TS44490-0050 TS44490-0250
4-Carbamoylphenylboronic acid 97%
4-Carbamoylphenylboronic acid
Formula: C₇H₈BNO₃
Melting Pt: 242…252 °C
Storage Temperature: Ambient
MDL Number: MFCD03411940
CAS Number: 123088-59-5

Order Now

Specification Test Results

Appearance White to tan Solid
Infrared spectrum Conforms
Additional info May contain varying amounts of anhydrides
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR