4-Hydroxy-6-methyl-2-pyrone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxy-6-methyl-2H-pyran-2-one, 3,5-Dihydroxysorbic acid d-lactone, 2-hydroxy-6-methyl-pyran-4-one

12156-0050 12156-0250
TS12156-0050EA 26 USD
TS12156-0050 TS12156-0250
4-Hydroxy-6-methyl-2-pyrone 98%
4-Hydroxy-6-methyl-2-pyrone
Formula: C₆H₆O₃
MW: 126.11 g/mol
Melting Pt: 185…189 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006641
CAS Number: 675-10-5

Order Now

Specification Test Results

Appearance Off-white to brown Powder
Infrared spectrum Conforms
Melting point 186 to 192 °C
HPLC ≥97.5 %
UV at 250 nm A: ≤0.01
Residue after evaporation ≤2 ppm
Acidity/alkalinity ≤0.0001 meq/g
Gradient analysis :
Absorbance largest eluted peak ≤0.002 A.U.
Filtered to 0.2 micrometer Filtered

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR