(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulphonate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-(+)-Glycidyl nosylate, (S)-Oxiran-2-ylmethyl 3-nitrobenzenesulphonate, Glycidyl (S)-(+)-3-nitrobenzenesulphonate

29642-0025 29642-0100 29642-0500
TS29642-0025EA 83 USD
TS29642-0025 TS29642-0100 TS29642-0500
(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulphonate 98%
(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulfonate
Formula: C₉H₉NO₆S
MW: 259.23 g/mol
Melting Pt: 61…65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064582
CAS Number: 115314-14-2

Order Now

Specification Test Results

Appearance Almost white to yellow-brown Crystalline powder
Infrared spectrum Conforms
Melting point 61 to 65 °C
GC ≥97.5 %
Specific optical rotation +21.0° to +25.0° (20 °C, 589 nm) (c=2, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR