Methyl-2-(hydroxymethyl)acrylate 85% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ) 0.1 wt.%
43590-0010
43590-0050
TS43590-0010EA
60
USD
TS43590-0010
TS43590-0050
Methyl-2-(hydroxymethyl)acrylate 85% stabilized
Methyl-2-(hydroxymethyl)acrylate
Formula:
C₅H₈O₃ Boiling Pt: 71…72 °C (2Torr) Density: 1.129 g/cm³ Flash Pt: 90 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD12545894 CAS Number: 15484-46-5 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
GC | ≥84.0 % |
NMR | consistent with structure |
Specific optical rotation | +8° to +11° (20°C, 589 nm) (c=1,CH3OH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...