1-Methyl-1H-pyrazole-4-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

44408-0010 44408-0050
TS44408-0010EA 92 USD
TS44408-0010 TS44408-0050
1-Methyl-1H-pyrazole-4-carboxylic acid 97%
1-Methyl-1H-pyrazole-4-carboxylic acid
Formula: C₅H₆N₂O₂
MW: 126.1100 g/mol
Melting Pt: 207…209 °C
Storage Temperature: Ambient
MDL Number: MFCD00159641
CAS Number: 5952-92-1

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR