3-(Boc-amino)propylbromide 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(tert-Butoxycarbonylamino)propyl bromide, 3-(Boc-amino)propyl bromide, tert-Butyl N-(3-Bromopropyl)carbamate, N-(3-Bromopropyl)carbamic acid tert-Butyl ester

39701-0010 39701-0050
TS39701-0010EA 194 USD
TS39701-0010 TS39701-0050
3-(Boc-amino)propylbromide 95%
3-(Boc-amino)propylbromide

Appearance: White Low melting solid

Formula: C8H16BrNO2
MW: 238.1221 g/mol
Melting Pt: 34…36.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683429
CAS Number: 83948-53-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White Low melting solid
Infrared spectrum Conforms
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR