tert-Butyl-3-oxocyclobutylcarbamate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Carbamic acid, N-(3-oxocyclobutyl)-, 1,1-dimethylethyl ester, 2-Methyl-2-propanyl (3-oxocyclobutyl)carbamate, tert-Butyl 3-oxocyclobutylcarbamate

44659-0010 44659-0050
TS44659-0010EA 245 USD
TS44659-0010 TS44659-0050
tert-Butyl-3-oxocyclobutylcarbamate 95%
tert-Butyl-3-oxocyclobutylcarbamate
Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD09751872
CAS Number: 154748-49-9

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or solid
Infrared spectrum Conforms
GC ≥94.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR