Pyrrolidine-3-carboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
H-DL-β-Pro-OH
45911-0010
45911-0050
TS45911-0010EA
89
USD
TS45911-0010
TS45911-0050
Pyrrolidine-3-carboxylic acid 98%
Pyrrolidine-3-carboxylic acid
Formula:
C5H9NO2 MW: 115.1305 g/mol Melting Pt: 182…184 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318699 CAS Number: 59378-87-9 |
Specification Test Results
Appearance | White to light orange Powder or solid |
Infrared spectrum | Conforms |
Elemental Analysis | ≥11.86 to ≤12.47 % Nitrogen |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...