tert-Butyl (2-bromoethyl)carbamate 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(BOC-amino)ethyl bromide

39894-0010 39894-0050
TS39894-0010EA 62 USD
TS39894-0010 TS39894-0050
tert-Butyl (2-bromoethyl)carbamate 96%
tert-Butyl (2-bromoethyl)carbamate

Appearance: Off-white to beige Low melting solid

Formula: C₇H₁₄BrNO₂
MW: 224.10
Melting Pt: 30…32 °C
Storage Temperature: Freezer
MDL Number: MFCD02683428
CAS Number: 39684-80-5

Order Now

Specification Test Results

Appearance Off-white to beige Low melting solid
Infrared spectrum Conforms
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR