4-Phenyl-1-butyne 97%

Supplier: Thermo Scientific Chemicals
Warning

44977-0050 44977-0250
TS44977-0050EA 106 USD
TS44977-0050 TS44977-0250
4-Phenyl-1-butyne 97%
4-Phenyl-1-butyne
Formula: C₁₀H₁₀
Boiling Pt: 190 °C
Density: 0.926 g/cm³
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00041667
CAS Number: 16520-62-0

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5210 to 1.5240 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR