(R)-(+)-2-Hydroxy-3-phenylpropionic acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: D-(+)-3-Phenyllactic acid, (R)-2-Hydroxy-3-phenylpropanoic acid, D(+)-Phenyllactic acid, D-​(+)​-​Phenyllactic acid

34458-0010 34458-0050
TS34458-0010EA 63 USD
TS34458-0010 TS34458-0050
(R)-(+)-2-Hydroxy-3-phenylpropionic acid 98%
(R)-(+)-2-Hydroxy-3-phenylpropionic acid
Formula: C₉H₁₀O₃
MW: 166.1739 g/mol
Melting Pt: 121…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00078062
CAS Number: 7326-19-4
EINECS: 230-803-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 121 to 125 °C
Specific optical rotation +17° to +20° (20 °C, 589 nm) (c=1, CH3OH)
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR