1,2-Ethanedisulphonic acid dihydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethane-1,2-disulphonic acid dihydrate

38480-0050 38480-0250
TS38480-0050EA 62 USD
TS38480-0050 TS38480-0250
1,2-Ethanedisulphonic acid dihydrate 98%
1,2-Ethanedisulphonic acid dihydrate
Formula: C₂H₆O₆S₂·2H₂O
MW: 226.2260 g/mol
Melting Pt: 109…113 °C
Storage Temperature: Ambient
MDL Number: MFCD26793415
CAS Number: 5982-56-9
EINECS: 203-732-0
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance Off-white to beige to grey to light brown Powder or crystals
Infrared spectrum Conforms
Water 14 to 19 % (K.F.)
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR