4-Cyano-3-fluorophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Cyano-3-fluorobenzeneboronic acid

44871-0010 44871-0050
TS44871-0010EA 57 USD
TS44871-0010 TS44871-0050
4-Cyano-3-fluorophenylboronic acid 97%
4-Cyano-3-fluorophenylboronic acid
Formula: C₇H₅BFNO₂
Melting Pt: 362…366 °C
Storage Temperature: Ambient
MDL Number: MFCD03411549
CAS Number: 843663-18-3

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR