Pyridazine-4-carboxylic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Pyridazinecarboxylic acid
39963-0010
39963-0050
TS39963-0010EA
106
USD
TS39963-0010
TS39963-0050
Pyridazine-4-carboxylic acid 95%
Pyridazine-4-carboxylic acid
Formula:
C₅H₄N₂O₂ MW: 124.10 g/mol Melting Pt: 244.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011576 CAS Number: 50681-25-9 |
Specification Test Results
Appearance | Off-white to brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...