Pyridazine-3-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

44672-0010 44672-0050
TS44672-0010EA 97 USD
TS44672-0010 TS44672-0050
Pyridazine-3-carboxylic acid 97%
Pyridazine-3-carboxylic acid
Formula: C₅H₄N₂O₂
Melting Pt: 190.8…194 °C
Storage Temperature: Ambient
MDL Number: MFCD01646333
CAS Number: 2164-61-6

Order Now

Specification Test Results

Appearance Off-white to yellow to light brown Powder or solid
Infrared spectrum Conforms
Water ≤1.5 %
NMR (1H-NMR) Consistent with structure
Titration with NaOH ≥96.0 to ≤104.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR