1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Isobutyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-Isobutylpyrazole-4-boronic acid, pinacol ester
43973-0010
43973-0050
TS43973-0010EA
82
USD
TS43973-0010
TS43973-0050
1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester 97%
1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester
Formula:
C₁₃H₂₃BN₂O₂ Boiling Pt: 248…249 °C (760 mmHg) Density: 1.003 g/cm³ Flash Pt: 48 °C Storage Temperature: Ambient |
MDL Number:
MFCD05663857 CAS Number: 827614-66-4 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...