Dimethyl-trans-cyclohexane-1,4-dicarboxylate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dimethyl trans-1,4-cyclohexanedicarboxylate, Dimethyl trans-1,4-Cyclohexanedicarboxylate

24079-0050 24079-0250
TS24079-0050EA 101 USD
TS24079-0050 TS24079-0250
Dimethyl-trans-cyclohexane-1,4-dicarboxylate 99+%
Dimethyl trans-cyclohexane-1,4-dicarboxylate
Formula: C₁₀H₁₆O₄
MW: 200.23 g/mol
Melting Pt: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00063917
CAS Number: 3399-22-2

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 67 to 70 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR