4-Methoxy-2-methylphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methoxy-2-methylbenzeneboronic acid, (4-Methoxy-2-methylphenyl)boronic acid

39385-0010 39385-0050
TS39385-0010EA 66 USD
TS39385-0010 TS39385-0050
4-Methoxy-2-methylphenylboronic acid 97%
4-Methoxy-2-methylphenylboronic acid
Formula: C₈H₁₁BO₃
MW: 165.99 g/mol
Melting Pt: 148…161 °C
Storage Temperature: Ambient
MDL Number: MFCD02684315
CAS Number: 208399-66-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline solid
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR