5-Pyrimidineboronic acid 97% may contain varying amounts of anhydride

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyrimidine-5-boronic acid, 5-Pyrimidylboronic acid, Pyrimidin-5-ylboronic acid, 5-Pyrimidinylboronic acid

45170-0250 45170-1000
TS45170-0250EA 282 USD
TS45170-0250 TS45170-1000
5-Pyrimidineboronic acid 97% may contain varying amounts of anhydride
5-Pyrimidineboronic acid
Formula: C₄H₅BN₂O₂
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03002366
CAS Number: 109299-78-7

Order Now

Specification Test Results

Appearance White to off-white Solid
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR