(S)-3-(Boc-amino)piperidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-3-Boc-aminopiperidine, (S)-tert-Butyl piperidin-3-ylcarbamate, tert-butyl N-[(3S)-piperidin-3-yl]carbamate
43653-0010
43653-0050
TS43653-0010EA
81
USD
TS43653-0010
TS43653-0050
(S)-3-(Boc-amino)piperidine 97%
(S)-3-(Boc-amino)piperidine
Formula:
C₁₀H₂₀N₂O₂ MW: 200.28 g/mol Melting Pt: 104.4…104.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD03093383 CAS Number: 216854-23-8 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Specific optical rotation | -2.5° to -3.5° (20°C, 589 nm) (c=1 in DMF) |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...