2-Sulphobenzoic cyclic anhydride 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-sulphobenzoic anhydride, 2-sulphobenzoic acid cyclic anhydride

18475-0250 18475-1000
TS18475-0250EA 96 USD
TS18475-0250 TS18475-1000
2-Sulphobenzoic cyclic anhydride 95%
2-Sulphobenzoic cyclic anhydride
Formula: C₇H₄O₄S
Boiling Pt: 184…186 °C (18 mmHg)
Melting Pt: 116…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00005879
CAS Number: 81-08-3
EINECS: 201-322-6

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Titration by coupling ≥94 %
HPLC ≥94 %
Specific optical rotation -17° to -21° (20°C, 589 nm) (0.5 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR