4-Biphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: [1,1'-Biphenyl]-4-ylboronic acid

39442-0010 39442-0050 39442-0250
TS39442-0010EA 39 USD
TS39442-0010 TS39442-0050 TS39442-0250
4-Biphenylboronic acid 97%
4-Biphenylboronic acid
Formula: C₁₂H₁₁BO₂
MW: 198.03 g/mol
Melting Pt: 241…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00093311
CAS Number: 5122-94-1

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR