3-Aminophenylboronic acid hemisulphate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3-Aminophenyl)boronic acid sulphate (2:1), 3-Aminobenzeneboronic acid hemisulphate

10430-0050 10430-0250
TS10430-0050EA 118 USD
TS10430-0050 TS10430-0250
3-Aminophenylboronic acid hemisulphate 98%
3-Aminophenylboronic acid hemisulphate
Formula: C₁₂H₁₈B₂N₂O₈S
MW: 371.97 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00013111
CAS Number: 66472-86-4

Order Now

Specification Test Results

Appearance White to beige to grey to light brown Powder or crystals
Infrared spectrum Conforms
Titration with NaOH 97.5 to 102.5 %
Fluoride (F) ≤50 ppm
Arsenic (As) ≤3 ppm
Barium (Ba) Passes test
Iron (Fe) ≤0.1 %
Lead (Pb) ≤3 ppm
Mercury (Hg) ≤0.5 ppm
Acid insoluble substances ≤0.2 %
Magnesium & Alkali Salts ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR