2-(Ethoxycarbonyl)phenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Ethoxycarbonyl)benzeneboronic acid, (2-(Ethoxycarbonyl)phenyl)boronic acid

39383-0050 39383-0250
TS39383-0050EA 87 USD
TS39383-0050 TS39383-0250
2-(Ethoxycarbonyl)phenylboronic acid 97%
2-(Ethoxycarbonyl)phenylboronic acid
Formula: C₉H₁₁BO₄
Melting Pt: 137…138 °C
Storage Temperature: Ambient
MDL Number: MFCD02179453
CAS Number: 380430-53-5

Order Now

Specification Test Results

Appearance White to off-white Solid
Infrared spectrum Conforms
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR