5-Indoleboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(1H-indol-5-yl)boronic acid, (1H-Indol-5-yl)boronic acid, 5-Indolylboronic acid
35946-0010
35946-1000
35946-5000
TS35946-0010EA
104
USD
TS35946-0010
TS35946-1000
TS35946-5000
5-Indoleboronic acid 97%
5-Indoleboronic acid
Formula:
C₈H₈BNO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD01319013 CAS Number: 144104-59-6 |
Specification Test Results
Appearance | Light yellow to beige Powder and/or chunks |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Titration with NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...