Monomethyl phthalate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Methoxycarbonyl)benzoic acid, Methyl hydrogen phthalate
37488-0250
37488-1000
TS37488-0250EA
130
USD
TS37488-0250
TS37488-1000
Monomethyl phthalate 98%
Monomethyl phthalate
Formula:
C₉H₈O₄ MW: 180.16 g/mol Melting Pt: 81…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002466 CAS Number: 4376-18-5 EINECS: 224-476-6 |
Specification Test Results
Appearance | White to off-white Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 81 to 86 °C |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...