Methyl-3,3-dimethoxypropionate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3,3-dimethoxypropanoate, Methyl 3,3-dimethoxypropionate

26434-0050 26434-0250
TS26434-0050EA 20 USD
TS26434-0050 TS26434-0250
Methyl-3,3-dimethoxypropionate 99%
Methyl 3,3-dimethoxypropionate
Formula: C₆H₁₂O₄
MW: 148.16 g/mol
Boiling Pt: 166 °C (760 mmHg)
Density: 1.06 g/cm³
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00010650
CAS Number: 7424-91-1
EINECS: 231-055-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4090 to 1.4110 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR