Ethyl-α-bromophenylacetate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethyl alpha-bromophenylacetate
43806-0050
43806-0250
43806-1000
TS43806-0050EA
49
USD
TS43806-0050
TS43806-0250
TS43806-1000
Ethyl-α-bromophenylacetate 97%
Ethyl α-bromophenylacetate
Formula:
C₁₀H₁₁BrO₂ Boiling Pt: 89…92 °C (1.2hPa) Density: 1.389 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013536 CAS Number: 2882-19-1 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5370 to 1.5420 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...