2-Acrylamido-2-methylpropanesulphonic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Acrylamido-2-methyl-1-propanesulphonic acid
40029-0050
40029-0500
TS40029-0050EA
25
USD
TS40029-0050
TS40029-0500
TS40029-2500
2-Acrylamido-2-methylpropanesulphonic acid 97%
2-Acrylamido-2-methylpropanesulphonic acid
Formula:
C₇H₁₃NO₄S MW: 207.25 g/mol Melting Pt: 195 °C (dec.) Flash Pt: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007522 CAS Number: 15214-89-8 EINECS: 239-268-0 |
Specification Test Results
Appearance | White Powder or chips and/or chunks |
Infrared spectrum | Conforms |
Titration with NaOH | ≥96.0 % |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...