Sodium glycolate 97%

Supplier: Thermo Scientific Chemicals

35157-0250 35157-1000
TS35157-0250EA 32 USD
TS35157-0250 TS35157-1000
Sodium glycolate 97%
Sodium glycolate
Formula: C₂H₃NaO₃
Storage Temperature: Ambient
MDL Number: MFCD00065358
CAS Number: 2836-32-0
EINECS: 220-624-9

Order Now

Specification Test Results

Appearance White to cream Powder and/or lumps
Infrared spectrum Conforms
Titration with HClO4 96.0 to 104.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR