Methyl glycolate 98%
Supplier: Thermo Scientific Chemicals
41437-0250
41437-1000
TS41437-0250EA
110
USD
TS41437-0250
TS41437-1000
Methyl glycolate 98%
Methyl glycolate
Formula:
C₃H₆O₃ Boiling Pt: 52 °C (17 mmHg) Melting Pt: 149…151 °C Density: 1.184 g/cm³ Flash Pt: 67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004667 CAS Number: 96-35-5 EINECS: 202-502-7 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4150 to 1.4180 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...