3-Bromo-1-propylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromopropylboronic acid pinacol ester, 2-(3-Bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

43393-0010 43393-0050
TS43393-0010EA 49 USD
TS43393-0010 TS43393-0050
3-Bromo-1-propylboronic acid pinacol ester 97%
3-Bromo-1-propylboronic acid pinacol ester
Formula: C₉H₁₈BBrO₂
Boiling Pt: 61…64 °C (.4 mmHg)
Density: 1.177 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD10567053
CAS Number: 124215-44-7

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR