O-Methylisourea hemisulphate 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
O-Methylisouronium sulphate
23303-0250
23303-2500
TS23303-0250EA
72
USD
TS23303-0250
TS23303-2500
O-Methylisourea hemisulphate 94%
O-Methylisourea hemisulfate
Formula:
C₄H₁₄N₄O₆S MW: 246.24 g/mol Melting Pt: 147…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040594 CAS Number: 52328-05-9 EINECS: 257-851-8 |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration with NaOH | ≥92 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...