3-(Trifluoromethyl)-5-pyridineboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (5-(Trifluoromethyl)pyridin-3-yl)boronic acid, (5-Trifluoromethylpyridin-3-yl)boronic acid

43052-0010 43052-2500
TS43052-0010EA 788 USD
TS43052-0010 TS43052-2500
3-(Trifluoromethyl)-5-pyridineboronic acid 95%
3-(Trifluoromethyl)-5-pyridineboronic acid
Formula: C₆H₅BF₃NO₂
MW: 190.9156 g/mol
Melting Pt: 260…262.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09952041
CAS Number: 947533-51-9

Order Now

Specification Test Results

Appearance White to yellow Powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥94.0 %
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR