1-Phenylcyclohexanecarboxylic acid 95%

Supplier: Thermo Scientific Chemicals

17075-0100 17075-0500
TS17075-0100EA 84 USD
TS17075-0100 TS17075-0500
1-Phenylcyclohexanecarboxylic acid 95%
1-Phenylcyclohexanecarboxylic acid
Formula: C₁₃H₁₆O₂
Melting Pt: 119…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00037152
CAS Number: 1135-67-7
EINECS: 214-495-8

Order Now

Specification Test Results

Appearance Beige to brown Powder
Infrared spectrum Conforms
Melting point 119 to 124 °C
Titration with NaOH ≥94 %
Stabilizer Approx. 200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR