Methyl-1-cyclopentene-1-carboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 1-cyclopentene-1-carboxylate
39759-0010
39759-0100
TS39759-0010EA
46
USD
TS39759-0010
TS39759-0100
Methyl-1-cyclopentene-1-carboxylate 97%
Methyl 1-cyclopentene-1-carboxylate
Formula:
C₇H₁₀O₂ MW: 126.1531 g/mol Boiling Pt: 76…78 °C (9 mmHg) Flash Pt: 62 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00239506 CAS Number: 25662-28-6 |
Specification Test Results
Appearance | Colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...