Methyl cyclopropanecarboxylate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methylcyclopropanecarboxylate

12671-0250 12671-1000 12671-5000
TS12671-0250EA 44 USD
TS12671-0250 TS12671-1000 TS12671-5000
Methyl cyclopropanecarboxylate 98%
Methyl cyclopropanecarboxylate
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 119 °C (764 mmHg)
Density: 0.98 g/cm³
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00001280
CAS Number: 2868-37-3
EINECS: 220-690-9
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4190 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR