Methyl cyclopentanecarboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
45092-0050
45092-0250
TS45092-0050EA
91
USD
TS45092-0050
TS45092-0250
Methyl cyclopentanecarboxylate 97%
Methyl cyclopentanecarboxylate
Formula:
C₇H₁₂O₂ Flash Pt: 43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00019290 CAS Number: 4630-80-2 UN: 3272 ADR: 3,III |
Specification Test Results
Appearance | Colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...