Dibenzothiophene-4-boronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Dibenzothiopheneboronic acid

35908-0010 35908-0050
TS35908-0010EA 46 USD
TS35908-0010 TS35908-0050
Dibenzothiophene-4-boronic acid 95%
Dibenzothiophene-4-boronic acid
Formula: C₁₂H₉BO₂S
MW: 228.08 g/mol
Melting Pt: 285…290 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01318182
CAS Number: 108847-20-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR