Diethyl vinylphosphonate 97%

Supplier: Thermo Scientific Chemicals

15215-0100 15215-0250 15215-1000
TS15215-0100EA 154 USD
TS15215-0100 TS15215-0250 TS15215-1000
Diethyl vinylphosphonate 97%
Diethyl vinylphosphonate
Formula: C₆H₁₃O₃P
Boiling Pt: 202 °C (760 mmHg)
Density: 1.06 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009079
CAS Number: 682-30-4
EINECS: 211-663-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4280 to 1.4300 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR