Diethyl allylphosphonate 97%

Supplier: Thermo Scientific Chemicals

33611-0050 33611-0250
TS33611-0050EA 100 USD
TS33611-0050 TS33611-0250
Diethyl allylphosphonate 97%
Diethyl allylphosphonate
Formula: C₇H₁₅O₃P
MW: 178.1660 g/mol
Boiling Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00015134
CAS Number: 1067-87-4

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4325 to 1.4345 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR