2-Methylfuran-3-boronic acid pinacol ester 90%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-(2-methyl-3-furyl)-1,3,2-dioxaborolane

45152-0010 45152-0050
TS45152-0010EA 216 USD
TS45152-0010 TS45152-0050
2-Methylfuran-3-boronic acid pinacol ester 90%
2-Methylfuran-3-boronic acid pinacol ester
Formula: C₁₁H₁₇BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD09879905
CAS Number: 864776-02-3

Order Now

Specification Test Results

Appearance Colorless to orange Liquid
Infrared spectrum Conforms
GC ≥89.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR