Tetraethyl methylenebisphosphonate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetraethyl methylenediphosphonate, Tetraethyl methylenebis(phosphonate)

30882-0050 30882-0250
TS30882-0010EA 28.42 USD
TS30882-0010 TS30882-0050 TS30882-0250
Tetraethyl methylenebisphosphonate 97%
Tetraethyl methylenebisphosphonate
Formula: C₉H₂₂O₆P₂
MW: 288.22 g/mol
Boiling Pt: 171…174 °C (11 mmHg)
Density: 1.16 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00039887
CAS Number: 1660-94-2
EINECS: 216-764-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4440 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR