Dimethyl (2-oxopropyl)phosphonate 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Dimethyl acetylmethylphosphonate
17761-0050
17761-0250
17761-1000
TS17761-0050EA
109
USD
TS17761-0050
TS17761-0250
TS17761-1000
Dimethyl (2-oxopropyl)phosphonate 95%
Dimethyl (2-oxopropyl)phosphonate
Formula:
C₅H₁₁O₄P Boiling Pt: 76…79 °C (3 mmHg) Flash Pt: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008769 CAS Number: 4202-14-6 EINECS: 224-110-5 |
Specification Test Results
Appearance | Clear colorless to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4380 to 1.4410 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...