Diethyl (2-bromoethyl)phosphonate 97%

Supplier: Thermo Scientific Chemicals
Warning

11418-0050 11418-0250
TS11418-0050EA 68 USD
TS11418-0050 TS11418-0250
Diethyl (2-bromoethyl)phosphonate 97%
Diethyl (2-bromoethyl)phosphonate
Formula: C₆H₁₄BrO₃P
Boiling Pt: 75 °C (1 mmHg)
Density: 1.27 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000239
CAS Number: 5324-30-1
EINECS: 226-193-3

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4590 to 1.4610 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR