tert-Butyl (diethylphosphono)acetate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-Butyl 2-(diethoxyphosphoryl)acetate, Diethyl(2-tert-butoxycarbonylmethyl)phosphonate

39775-0010 39775-0050
TS39775-0010EA 36 USD
TS39775-0010 TS39775-0050 TS39775-0250
tert-Butyl (diethylphosphono)acetate 95%
tert-Butyl (diethylphosphono)acetate
Formula: C₁₀H₂₁O₅P
MW: 252.25 g/mol
Density: 1.074 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00075414
CAS Number: 27784-76-5

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR