Allylboronic acid pinacol ester 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Allyl-4,4,5,5-Tetramethyl-1,3,2-Dioxaborolane

39513-0010 39513-0050
TS39513-0010EA 48 USD
TS39513-0010 TS39513-0050
Allylboronic acid pinacol ester 98%
Allylboronic acid pinacol ester
Formula: C₉H₁₇BO₂
Boiling Pt: 50…53 °C (5 mmHg)
Flash Pt: 46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013347
CAS Number: 72824-04-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR