N,N-Diphenylcarbamoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diphenylcarbamyl chloride, Diphenylcarbamoyl chloride

16714-0250 16714-1000 16714-5000
TS16714-0250EA 56 USD
TS16714-0250 TS16714-1000 TS16714-5000
N,N-Diphenylcarbamoyl chloride 98%
N,N-Diphenylcarbamoyl chloride
Formula: C₁₃H₁₀ClNO
MW: 231.68 g/mol
Melting Pt: 81…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00000633
CAS Number: 83-01-2
EINECS: 201-450-2
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance Beige to grey-green Crystalline powder
Infrared spectrum Conforms
Melting point 81 to 85 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR