Diethyl (cyanomethyl)phosphonate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
11427-0100
11427-0500
11427-2500
TS11427-0100EA
63.7
USD
TS11427-0100
TS11427-0500
TS11427-2500
Diethyl (cyanomethyl)phosphonate 96%
Diethyl (cyanomethyl)phosphonate
Formula:
C₆H₁₂NO₃P Boiling Pt: 101…102 °C (.4 mmHg) Density: 1.09 g/cm³ Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001893 CAS Number: 2537-48-6 EINECS: 219-806-0 UN: 3278 ADR: 6.1,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4312 to 1.4332 (20°C, 589 nm) |
GC | ≥95 % |
Additional info | Cr2O3: 0.00005 % |
Particle size | 205 µm (D50) Typical |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...